Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Lactate Dehydrogenase Reagents for Hitachi™ Analyzers

Catalog No. 23666347 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
4 x 100, 8 x 21mL
4 x 50mL R1, 4 x 21mL R2
5 x 100 R2mL
5 x 600 R1mL
Formulation:
NAD 7.5mM, L-Lactate 55mM, Buffer pH 8.95, non-reactive Stabilizers and Sodium Azide 0.1% as preservative
Sodium Azide 0.095%
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
23-666-347 4 x 100, 8 x 21mL Sodium Azide 0.095%
23-666-344 5 x 100 R2mL Sodium Azide 0.095%
23-666-343 5 x 600 R1mL Sodium Azide 0.095%
23-666-346 4 x 50mL R1, 4 x 21mL R2 NAD 7.5mM, L-Lactate 55mM, Buffer pH 8.95, non-reactive Stabilizers and Sodium Azide 0.1% as preservative
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 23-666-347 Supplier Pointe Scientific Supplier No. HL956568

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Reagent 1 and Reagent 2 when combined: NAD 7.5mM; L-Lactate 55mM; Buffer (pH 8.95); Non-reactive stabilizers; Sodium azide 0.1%

Quantify lactate dehydrogenase activity easily with system specific reagents

  • Method: Modified Wacker/Tris, Kinetic
  • Format: Liquid (ready to use)
  • Linearity: 1000 U/L
  • Expected Values: 50 to 166 U/L (Male, 30°C); 60 to 132 U/L (Female, 30°C)

Applications:

For the quantitative kinetic determination of lactate dehydrogenase activity in serum on Hitachi analyzers

Specifications

Certifications/Compliance CE Marked
Type Lactate Dehydrogenase Reagent Kit
Formulation Sodium Azide 0.095%
For Use With (Application) Quantitative kinetic determination of Lactate Dehydrogenase activity in Serum
Quantity 4 x 100, 8 x 21mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.