Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Liquid-Stable CO2 Reagents

Catalog No. 22666300 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
1 x 120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-666-300 1 x 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-666-300 Supplier Pointe Scientific Supplier No. C7502120
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Certifications/Compliance CE Marked
Type Carbon Dioxide Reagent
Formulation PEP 6mM, Magnesium ions 10mM, NADH analog, MDH, PEP 6mM, Magnesium Ions 10mM, NADH analog, MDH, reactive Stabilizers with Surfactants and preservative
For Use With (Application) Quantitative determination of Carbon Dioxide in Serum
Quantity 1 x 120 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.