Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Microalbumin Liquid Reagents

Catalog No. 23666372 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
4 x 20mL R1, 2 x 10mL R2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-666-372 4 x 20mL R1, 2 x 10mL R2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-666-372 Supplier Pointe Scientific Supplier No. M7562100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

  • Buffer Reagent: Tris (hydroxymethyl) aminomethane100mM; pH 7.6
  • Antiserum Reagent: Anti-human albumin, goat antiserum 20%; Tris (hydroxymethyl) aminomethane 100mM; pH 7.6

Requires

Microalbumin Multi-Calibrator Set

Quantify low levels of albumin in urine

  • Method: Immunoturbidimetric
  • Format: Liquid (ready to use)
  • Wavelength: 340/700nm
  • Linearity: 0.5 to 30.0mg/dL
  • Expected Values: 30 to 300mg/dL (24 hr.)

For the quantitative determination of low levels of albumin in urine

Specifications

Certifications/Compliance CE Marked
Type Microalbumin Reagent Set
Formulation Buffer reagent: Tris (hydroxymethyl) Aminomethane 100mM, antiserum reagent: Anti-Human Albumin 20%, Goat antiserum, Tris (hydroxymethyl) Aminomethane 100mM
For Use With (Application) Quantitative determination of low levels of Albumin in Urine by immunoturbidimetric assay
Includes Buffer reagent, Antiserum reagent
Quantity 4 x 20mL R1, 2 x 10mL R2
Wavelength 340/700 nm
Form Liquid

For in vitro diagnostic use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.