Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Phosphorus Reagents

Catalog No. 23666385 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
1 x 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-666-385 1 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-666-385 Supplier Pointe Scientific Supplier No. P7516500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Ammonium molybdate 0.48mM; Sulfuric acid 220mM; surfactant

Quantify inorganic phosphorus in serum

  • Method: Phosphomolybdate, Endpoint
  • Format: Liquid (ready to use)
  • Wavelength: 340nm
  • Linearity: 12mg/dL
  • Expected Values: 2.5 to 4.8mg/dL

For the quantitative determination of Inorganic Phosphorus in serum

Specifications

Certifications/Compliance CE Marked
Type Inorganic Phosphorus Reagent Set
Formulation Ammonium Molybdate 0.48mM, Sulfuric Acid 220mM with Surfactant
For Use With (Application) Quantitative determination of Inorganic Phosphorus in Serum
Quantity 1 x 500 mL
Wavelength 340 nm
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.