Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Phosphorus Reagents for Hitachi™ Analyzers

Catalog No. 23666381 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
5 x 300 R2mL
5 x 600 R1mL
8 x 50mL R1, 4 x 50mL R2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-666-381 5 x 300 R2mL
23-666-380 5 x 600 R1mL
23-666-377 8 x 50mL R1, 4 x 50mL R2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-666-381 Supplier Pointe Scientific Supplier No. HP416R2

missing translation for 'validMainframeMsg'

Includes

Reagent 1 and Reagent 2 when combined: Ammonium molybdate >0.48mM; Sulfuric acid <220mM; surfactant

Requires

Calibrator/standard

Quantify inorganic phosphorus easily with system specific reagents

  • Method: Phosphomolybdate, Endpoint
  • Format: Liquid (ready to use)
  • Linearity: 12mg/dL
  • Expected Values: 2.5 to 4.8mg/dL

Applications:

For the quantitative determination of inorganic phosphorus in serum on Hitachi analyzers

Specifications

Certifications/Compliance CE Marked
Type Phosphorus R2
Formulation Ammonium Molybdate >0.48mM, Sulfuric Acid <220mM with Surfactant
For Use With (Application) Quantitative determination of inorganic Phosphorus in Serum
Quantity 5 x 300 R2mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.