Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Triglycerides Liquid Reagents

Catalog No. 23666411 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
1 x 120 mL
1 x 500 mL
1 x 1 L
Type:
Triglyceride Reagent Kit
Triglyceride Reagent Set
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
23-666-411 1 x 1 L Triglyceride Reagent Set
23-666-410 1 x 120 mL Triglyceride Reagent Set
23-666-412 1 x 500 mL Triglyceride Reagent Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-666-411 Supplier Pointe Scientific Supplier No. T75321L
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

Certifications/Compliance CE Marked
Type Triglyceride Reagent Set
For Use With (Application) Quantitative determination of Triglyceride in Serum or Plasma
Quantity 1 x 1 L
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.