Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Polarizing Film
SDPSCI_ED

Catalog No. S15923 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Dimensions (L x W):
3 x 3 in.
6 x 6 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
S15923 3 x 3 in.
S15920 6 x 6 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S15923 Supplier United Scientific™ Supplier No. PLF030
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

An effective yet economical tool for demonstrating polarization of light.

  • Pack of two film squares
  • Effective way to demonstrate polarization of light.

Specifications

Product Type Polarizing Film
Dimensions (L x W) 3 x 3 in.

Warning: Cancer and reproductive harm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.