Learn More
Description
- Designed to withstand temperature from -196 to 121°C
- Printed barcode and white writing surface aid in specimen identification
- Certified at 95 kPa to provide a leakproof seal
Cryogenic storage vials feature a co-molded screw cap that combines polyethylene (PE) with a thermoplastic elastomer (TPE), eliminating silicone O-rings for a safer sample storage environment. Cap design provides a secure seal on the inside of the vial, makes gripping the outside of the cap easier, and works well with automatic decapper machines. Caps are offered with external or internal threaded configurations. Multicolored cap inserts (not included) are available to aid in visual identification.
Vials are available with round or self-standing skirted bottoms. A special locking system on the bottom of each skirted vial interlocks to a cryogenic rack (not included) for easy one-handed manipulation.
The vials are manufactured in a ISO Class 7 cleanroom in accordance with FDA, GMP, and ISO quality standards. Sterilized by gamma irradiation, they are packaged in resealable bags of 50. They are also certified to be DNase, RNase, pyrogen, ATP and human DNA free.
Do not use vials for storage in the liquid phaes of liquid nitrogen: this may cause entrapment of liquified nitrogen inside the vial and lead to pressure build-up,resulting in possible explosion or biohazard release.
Specifications
Specifications
| Color | Clear |
| Diameter (Metric) Outer | 12.5 mm |
| Material | Polyethylene |
| Pre-slit | No |
| Sterility | Sterile |
| Volume (Metric) | 4 mL |
| Height (English) | 2.8 in. |
| Temperature Range (Metric) | -196°C to +121°C |
| Thread Style | Internal |
| Base Style | Round |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.