Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POLE4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597283
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597283 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597283 Supplier Invitrogen™ Supplier No. PA597283
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAAAAAAGSG TPREEEGPAG EAAASQPQAP TSVPGARLSR LPLARVKALV KADPDVTLAG QEAIFILARA AELFVETIAK DAYCCAQQGK RKTLQRRDLD NAIEAVDEFA FLEGTLD; Positive Samples: HeLa, 293T, HepG2, MCF-7, HL-60; Cellular Location: Nucleus.

POLE4 is a histone-fold protein that interacts with other histone-fold proteins to bind DNA in a sequence-independent manner. These histone-fold protein dimers combine within larger enzymatic complexes for DNA transcription, replication, and packaging.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLE4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.04 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene POLE4
Gene Accession No. Q9NR33
Gene Alias 2400007P05Rik; 5830430F06Rik; DNA polymerase epsilon 4, accessory subunit; DNA polymerase epsilon p12 subunit; DNA polymerase epsilon subunit 4; DNA polymerase epsilon subunit p12; DNA polymerase II subunit 4; DNA-directed DNA polymerase epsilon 4; p12; Pole4; polymerase (DNA) epsilon 4, accessory subunit; polymerase (DNA-directed), epsilon 4 (p12 subunit); polymerase (DNA-directed), epsilon 4, accessory subunit; YHHQ1
Gene Symbols POLE4
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56655
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.