Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POLG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521314 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521314 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521314 Supplier Invitrogen™ Supplier No. PA521314
  • $624.00
    $494.65 / Each of 1

    Save $129.35

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Recommended positive controls: MCF-7, MDA-MB-231, Rat testis. Predicted reactivity: Mouse (91%), Rat (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Mitochondrial DNA polymerase is heterotrimeric, consisting of a homodimer of accessory subunits plus a catalytic subunit. The protein encoded by this gene is the catalytic subunit of mitochondrial DNA polymerase. The encoded protein contains a polyglutamine tract near its N-terminus that may be polymorphic. Defects in this gene are a cause of progressive external ophthalmoplegia with mitochondrial DNA deletions 1 (PEOA1), sensory ataxic neuropathy dysarthria and ophthalmoparesis (SANDO), Alpers-Huttenlocher syndrome (AHS), and mitochondrial neurogastrointestinal encephalopathy syndrome (MNGIE). Two transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLG
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.67 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene POLG
Gene Accession No. P54098, Q9QYV8
Gene Alias AA409516; DNA polymerase gamma; DNA polymerase gamma subunit 1 (Mitochondrial DNA polymerase catalytic subunit) (PolG-alpha); DNA polymerase gamma, catalytic subunit; DNA polymerase subunit gamma-1; DNA-directed DNA polymerase gamma; FLJ27114; MDP1; Mip1; MIRAS; Mitochondrial DNA polymerase catalytic subunit; mitochondrial DNA polymerase gamma; mitochondrial DNA polymerase gamma catalytic subunit; mitochondrial DNA polymerase-gamma; mitochondrial polymerase gamma; MTDPS4A; MTDPS4B; PEO; Pol gamma; Polg; POLG1; POLGA; PolG-alpha; polymerase (DNA directed), gamma; polymerase (DNA) gamma, catalytic subunit; polymerase, gamma; SANDO; SCAE; truncated mitochondrial DNA polymerase gamma catalytic subunit
Gene Symbols POLG
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human DNA polymerase gamma. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5428, 85472
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.