Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POLI Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566707
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566707 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566707 Supplier Invitrogen™ Supplier No. PA566707
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PKDKETNRDFL PSGRIESTRT RESPLDTTNF SKEKDINEFP LCSLPEGVDQ EVFKQLPVDI QEEILSGKSR EKFQGKGSVS CPL Highest antigen sequence identity to the following orthologs - mouse 74%, rat 73%.

Error-prone DNA polymerase specifically involved in DNA repair. Plays an important role in translesion synthesis, where the normal high-fidelity DNA polymerases cannot proceed and DNA synthesis stalls. Favors Hoogsteen base-pairing in the active site. Inserts the correct base with high-fidelity opposite an adenosine template. Exhibits low fidelity and efficiency opposite a thymidine template, where it will preferentially insert guanosine. May play a role in hypermutation of immunoglobulin genes. Forms a Schiff base with 5'-deoxyribose phosphate at abasic sites, but may not have lyase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLI
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Poli
Gene Accession No. Q9UNA4
Gene Alias DNA polymerase iota; eta2; Poli; polymerase (DNA directed) iota; polymerase (DNA directed), iota; polymerase (DNA) iota; polymerase (DNA-directed), iota; RAD 30B; RAD30; RAD30 homolog B; RAD30B; RAD3OB
Gene Symbols Poli
Host Species Rabbit
Immunogen Recombinant Human POLI. Recombinant protein control fragment (Product #RP-107359).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11201
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.