Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Polishers for Milli-Q™ IQ 7000 Ultrapure Water System

Catalog No. V0CPAK0A1 Shop All MilliporeSigma Products
Change view
Click to view available options
Type:
Biopak™ Polisher
EDS-Pak™ Polisher
LC-Pak™ Polisher
VOC-Pak™ Polisher
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
V0CPAK0A1 VOC-Pak™ Polisher
LCPAK00A1 LC-Pak™ Polisher
CDUFBI0A1 Biopak™ Polisher
EDSPAK0A1 EDS-Pak™ Polisher
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. V0CPAK0A1 Supplier MilliporeSigma™ Supplier No. V0CPAK0A1
Add to Cart
Add to Cart

Our Lab Water purification system portfolio offers a broad range of pure and ultrapure water purification systems and services designed for scientists working in pharmaceutical, clinical, academic, industrial, research, and government laboratories — in both validated and non-validated environments.

Production of volatile organic compound-free water at the point of dispense of the Milli-Q™ IQ 7000 system.
TRUSTED_SUSTAINABILITY

Specifications

Description Delivered hermetically sealed
For Use With (Equipment) Milli-Q IQ 7000 Ultrapure Water System
Type VOC-Pak™ Polisher
For Use With (Application) Production of volatile organic compound-free water at the point of dispense of the Milli-Q IQ 7000 system
Purity Meter Included No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.