Learn More
Essendant Stainless Steel Cleaner and Polish, Lemon, 18 oz Aerosol Spray, 12/Carton
Supplier:  Essendant BWK347ACT
Encompass_Preferred
Powerful and reliable cleaner to wash and polish stainless steel, chrome, copper, aluminum, brass, and other metal surfaces. The water-in-oil emulsion formula means the cleaning product acts like a solvent (oil-based) without the oiliness or flammability of typical oil-based solvents. Pleasantly fragranced polish cuts through oil and grease, leaving a protective shine without streaks or films. Contains no chlorinated solvents or ozone depletion causing agents.
Catalog No. 27-934-39
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.