Learn More
Invitrogen™ POLL Polyclonal Antibody

Description
The antibody detects endogenous levels of total DNA Polymerase lambda protein.
Specifications
Specifications
| Antigen | POLL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Poll |
| Gene Accession No. | Q5RKI3, Q9QXE2, Q9UGP5 |
| Gene Alias | 1110003P06Rik; AV007317; BETAN; DNA polymerase beta-2; DNA polymerase beta-N; DNA polymerase kappa; DNA polymerase lambda; DNA-directed DNA polymerase lambda; EC 2.7.7.7; FLJ46002; OTTHUMP00000020323; Pol beta2; Pol Lambda; Polk; POLKAPPA; POL-KAPPA; Poll; polymerase (DNA directed), lambda; polymerase (DNA) lambda; RP11-529I10.1 |
| Gene Symbols | Poll |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.