Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poloxamer 181, Surfactant, Spectrum™ Chemical
SDP

Catalog No. 18605262 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
2.5 L
20 L
200 L
Packaging:
Drum
Glass Bottle
Poly Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-262 2.5 L Glass Bottle
18-605-263 20 L Poly Drum
18-605-264 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-605-262 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P11622.5LTGL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

P1162, 9003-11-6, HO(C2H4O)a(C3H6O)b(C2H4O)aH

Spectrum™ Chemical Poloxamer 181, Surfactant is a nonionic triblock copolymer. It is made up of a main hydrophobic chain of polyoxypropylene bordered on each side by two hydrophilic chains of polyoxyethylene. These surfactants are commonly used in industrial applications, cosmetics, and pharmaceuticals. They have also been evaluated for various drug delivery applications and were shown to sensitize drug resistant cancers to chemotherapy. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 9003-11-6
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
InChI Key RVGRUAULSDPKGF-UHFFFAOYNA-N
IUPAC Name 2-methyloxirane; oxirane
SMILES C1CO1.CC1CO1

Specifications

pH 5.0 to 7.5
CAS Max % 1
Quantity 2.5 L
Grade Reagent
Packaging Glass Bottle
Water 0.004
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.