Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poloxamer 182, Spectrum™ Chemical
SDP

Catalog No. 18605266 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
2.5 L
20 L
55 gal.
Packaging:
Drum
Glass Bottle
Steel Can
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-266 2.5 L Glass Bottle
18-605-267 20 L Steel Can
18-605-268 55 gal. Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-605-266 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P18132.5LTGL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

P1813, 9003-11-6, HO(C2H4O)a(C3H6O)b(C2H4O)aH

Spectrum™ Chemical Poloxamer 182

Chemical Identifiers

CAS 9003-11-6
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
InChI Key RVGRUAULSDPKGF-UHFFFAOYNA-N
IUPAC Name 2-methyloxirane; oxirane
SMILES C1CO1.CC1CO1

Specifications

pH 5.0 to 7.5
CAS Max % 1
Quantity 2.5 L
Grade Reagent
Packaging Glass Bottle
Water 0.004
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.