Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ Poloxamer 188, 10% Solution

Catalog No. MIR6230 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR6230 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MIR6230 Supplier Mirus Bio™ Supplier No. MIR 6230
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Poloxamer 188 is added to batch mammalian cell cultures to reduce foaming.

  • Clear, colorless solution
  • Molecular Formula: (C3H6O.C2H4O)X
  • CAS number: 9003-11-6
  • Non-ionic detergent
  • Improve resistance of cell membranes to hydrodynamic shearing
  • 0.1 μm sterile-filtered provided at concentration of 10%
  • Recommended working concentration: 0.1 to 0.3% (10 to 30 mL/L) of culture medium

Specifications

Content And Storage Store Poloxamer 188 Solution at 15° to 30°C.
Form Clear, colorless solution
Product Type Poloxamer 188
Sterility Sterile-filtered
Sterilization Method 0.1 μm Sterile-filtered
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.