Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POLR1E Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103797
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103797 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103797 Supplier Invitrogen™ Supplier No. PA5103797
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RPA49.

DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Component of RNA polymerase I which synthesizes ribosomal RNA precursors (PubMed:24207024). Appears to be involved in the formation of the initiation complex at the promoter by mediating the interaction between Pol I and UBTF/UBF (PubMed:24207024). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLR1E
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Polr1e
Gene Accession No. Q8K202, Q9GZS1
Gene Alias 53kDa; AU042259; D030019D19Rik; DNA-directed RNA polymerase I subunit E; DNA-directed RNA polymerase I subunit RPA49; FLJ13390; FLJ13970; hypothetical protein LOC511587; Paf53; Polr1e; polymerase (RNA) I associated factor 1; polymerase (RNA) I polypeptide E; polymerase (RNA) I polypeptide E, 53kDa; polymerase (RNA) I subunit E; PRAF1; RGD1565773; RNA polymerase I associated factor (Paf53); RNA polymerase I associated factor 53; RNA polymerase I subunit A49; RNA polymerase I subunit E; RNA polymerase I-associated factor 1; RNA polymerase I-associated factor 53; RP11-405L18.3
Gene Symbols Polr1e
Host Species Rabbit
Immunogen A synthesized peptide derived from human POLR1E(Accession Q9GZS1), corresponding to amino acid residues K109-G159.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64424, 64425
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.