Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POLR2H Monoclonal Antibody (OTI1H2), TrueMAB™, OriGene
SDP

Catalog No. 501683602
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-3602 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-3602 Supplier OriGene Supplier No. TA810716
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Eukaryotes produce 3 distinct classes of RNA polymerase, Pol I, II and III. Each polymerase is responsible for the synthesis of a different class of RNA. RNA polymerase I (Pol I) transcribes the rRNA (ribosomal RNA) genes for the precursor of the 28S, 18S, and 5.8S molecules of the ribosome. RNA polymerase II transcribes protein-encoding genes into mRNA (messenger RNA) and snRNA (small nuclear RNA) genes into snRNAs that influence the processing of other classes of RNA. RNA polymerase III (Pol III) transcribes the 5S rRNA genes and all of the tRNA (transfer RNA) genes. Each class of RNA polymerase is assembled from 9 to 15 different polypeptides. The RPB6 and RPB8 subunits are shared by all 3 RNA polymerases.

Specifications

Antigen POLR2H
Applications Western Blot
Classification Monoclonal
Clone OTI1H2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene POLR2H
Gene Accession No. P52434
Gene Alias RPABC3, RPB17, RPB8
Gene Symbols POLR2H
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 2-150 of human POLR2H produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5437
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.