Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Poly(A) Tail-Length Assay Kit

Catalog No. 764551KT
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-455-1KT 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 76-455-1KT Supplier Invitrogen™ Supplier No. 764551KT
  • $906.00
    $806.65 / Each of 1

    Save $99.35

    11% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This kit utilizes a novel G/I extension technique which preserves the 3' end of RNAs and ensures accurate measurement of poly(A) tail-length. And, all reaction components have been optimized so you get quality data quickly.

Poly adenylation of eukaryotic mRNAs plays a critical role in RNA metabolism, including stability, enhanced translation, nuclear export, and miRNA mediated gene regulation. The Poly(A) Tail-Length Assay Kit allows you to quickly measure the poly(A) tails of multiple mRNAs in only three hours. This kit utilizes a novel G/I extension technique that preserves the 3' end of RNAs and ensures accurate measurement of poly(A) tail-length. And all reaction components have been optimized so you get quality data quickly.

Features

  • Measure the poly(A) tail of multiple RNAs in under 3 hrs using common lab equipment
  • Accurate measurement of poly(A) tail length compared to other assays
  • One kit includes all necessary reagents for 5 G/I tailing, 20 RT, and 80 PCR reactions

Technology

The Poly(A) Tail-Length Assay Kit uses four key steps to enable poly(A) tail-length determination (see figure).

  • Step 1—poly(A) polymerase adds a limited number of guanosine and inosine residues to the 3'-ends of poly(A)-containing RNAs.
  • Step 2—the tailed-RNAs are converted to DNA through reverse transcription using the newly added G/I tails as the priming sites.
  • Step 3—PCR amplification products are generated using two primer sets. A gene-specific forward and reverse primer set designed upstream of the polyadenylation site (e.g., the 3'-UTR) is produced as a control for the gene-of-interest. The second set of primers uses the gene-specific forward primer and the universal reverse primer provided with the kit to generate a product that includes the poly(A) tails of the gene-of-interest.
  • Step 4—the PCR products are separated on an agarose or polyacrylamide gel (see figure).

Selected references

  • Parker, R. and Song, H. (2004) Nat Struct Mol Biol. 11 121-127.
  • Brodersen, D.E. et al. (2008) Biochim Biophys Acta. 1779, 532-537.
  • Wu, L., Fan J. and Belasco, J.G. (2006) Proc Natl Acad Sci U S A. 103, 4034-4039.
  • Izaurralde, E. et al. (2009) RNA 15, 21-32.
  • Martin, G. and Keller, W. (1998) RNA 4, 226-230.
  • Gauss-Müller, V. et al. (2001) Nucleic Acids Res. 29 E57-7
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Sufficient reagents for 5 G/I tailing, 20 reverse transcription, and 80 PCR reactions.

HeLa Total RNA and Control Human Actin PCR Forward Primer to verify components and protocol.

Store reagents at -15°C to -30°C in a non-frost-free freezer.
Store HeLa Total RNA at -80°C.
Shipping Condition Dry Ice
Polymerase Poly(A) RNA Polymerase
For Use With (Application) Poly(A) tail-length detection
Starting Material Total Eukaryotic RNA (100 ng to 2 μg)
Quantity 1 kit
Concentration 1.25 U/μL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.