Learn More
Sigma Organic Chemistry Poly(acrylic acid) | 5G | 2594322 | MFCD00084394
Supplier: Sigma Organic Chemistry 1812855G

Poly(acrylic acid), 5G
About This Item:
MDL Number: MFCD00084394
UNSPSC Code: 12162002
Storage: room temp
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.