Learn More
Akina Inc Poly(lactide-co-glycolide)-b-poly(ethylene glycol)-maleimide (per 100mg)
SDP
Supplier:  Akina Inc AI052100MG
Encompass
PLGA-PEG-Mal LG 75:25 (Mw ~60000:3400 Da) Poly(lactide-co-glycolide)-b-poly(ethylene glycol)-maleimide (per 100mg)
Catalog No. NC2827943