Learn More
StatLab POLY BATH 30ML
SDP
Supplier:  StatLab TAPBA
Encompass_Preferred
Poly Bath; 30mL; Master Tech; Adhesive; Binds sections to glass by interaction of heat in drying process with tissue anionic sites; Virtually eliminates background staining; Just by adding five drops to waterbath
Catalog No. NC9330783
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.