Learn More
CritiCore Protective Wear Poly/Cotton Coverall without Pockets in White
Catalog No. 17012891
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-012-891 3X-Large
17-012-885 X-Small
17-012-886 Small
17-012-887 Medium
17-012-888 Large
17-012-889 X-Large
17-012-890 2X-Large
17-012-892 4X-Large
17-012-893 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 17-012-891 Supplier CritiCore Protective Wear Supplier No. 13C86643PC45013XLWHT
Add to Cart
Add to Cart
This item is not returnable. View return policy
This poly/cotton coverall without pockets in white is constructed with 65/35 poly/cotton, offering the durability of polyester and the breathability of cotton. Lightweight and versatile, with a snap-front enclosure.
  • 65/35 Poly/Cotton
  • Snap Front Enclosure
  • Plain Hem Sleeve and Pant
  • No pockets, One pen tab
  • Mandarin Collar Primary Applications: Food Processing, Food Service, and Lab Environments
Specifications
Product Type Cleanroom Coveralls
Size 3X-Large
Gender Unisex
Material Polyester, Carbon
Color White
Disposable Reusable
Ankle Style Elastic
Cuff Style Open
Neck Style Collared
Closure Type Snap
Seam Type Stitched
Sterility Non-sterile
Resistance Particle