Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(diallyldimethylammonium chloride) solution

Catalog No. 40902225ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-902-225ML 25 mL
40-902-24L 4 L
40-902-21L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 40-902-225ML Supplier Sigma Aldrich Supplier No. 40902225ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly(diallyldimethylammonium chloride) solution (PDADMAC) is a cationic polyelectrolyte prepared by the radical polymerization of diallyldimethylammonium chloride (DADMAC) in presence of a catalyst in the form of an organic peroxide. It is widely utilized as a flocculant.PDADMAC can be used as an electrolytic solution in a variety of applications such as drug delivery, biomedical systems, and sensors, which include biosensors and chemical sensors.

Specifications

CAS 26062-79-3
Density 1.04 g/mL (at 25°C)
Refractive Index n20/D 1.375
Linear Formula (C8H16ClN)n
MDL Number MFCD00192409
Quantity 25 mL
Synonym PDADMAC
Molecular Weight (g/mol) 200,000-350,000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.