Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol)

Catalog No. 435465250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-546-5250ML 250 mL
43-546-51L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 43-546-5250ML Supplier Sigma Aldrich Supplier No. 435465250ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

PEG-PPG-PEG (Pluronic P123) symmetric triblock copolymer constitutes of poly(ethylene oxide)(PEO) and poly (propylene oxide) (PPO). The unique characteristic of PPO block exhibiting hydrophobicity at temperatures above 288K and solubility in water at temperatures below 288K lead to formation of micelle consisting of PEO-PPO-PEO triblock copolymers. Some studies report that the hydrophobic core contains PPO block and a hydrophilic corona is comprised of PEO block. In 30wt% aqueous solution Pluronic P123(R) forms a cubic gel phase.PEG-PPG-PEG is used in the preparation and coating of Fe-W-C(O) nanoparticles by sol gel technique. Also, employed as sacrificial templates to prepare ZnS nanospheres, nanoporous silica.Pluronic is a registered trademark of BASF

Specifications

Density 1.018 g/mL (at 25°C)
Refractive Index n20/D 1.465
MDL Number MFCD00082049
Quantity 250 mL
Synonym Pluronic™ P-123; PEG-PPG-PEG
Molecular Weight (g/mol) 5800
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.