Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(ethylene glycol) diglycidyl ether

Catalog No. 475696100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-569-6100ML 100 mL
47-569-6500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 47-569-6100ML Supplier Sigma Aldrich Supplier No. 475696100ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly(ethylene glycol) diglycidyl ether (PEGDGE) shows highly solubility in water. Hence, it easily undergoes hydrolysis followed by ring cleavage reaction in aqueous solution, yielding hydroxyl group. PEGDGE combines with proteins covalently or non-covalently. PEGDGE is widely used in chemical industries for cross linking and surface modifier.The high solubility of PEGDGE has been successfully employed to immobilize glucose oxidase, d-amino acid oxidase and glutamate oxidase. It may be used as a component for the development of microelectrode biosensors to detect hydrogen peroxide and nitric oxide.

Specifications

CAS 26403-72-5
Refractive Index n20/D 1.47
Linear Formula C3H5O2-(C2H4O)n-C3H5O
Quantity 100 mL
Synonym Diepoxy PEG; PEG diglycidyl ether; Polyoxyethylene bis(glycidyl ether)
Molecular Weight (g/mol) 500
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.