Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(ethylene glycol) methyl ether methacrylate solution

Catalog No. 4578761L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-787-61L 1 L
45-787-6250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-787-61L Supplier Sigma Aldrich Supplier No. 4578761L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Surface grafted polyethylene glycol (PEG) prevents protein absorption. Poly (ethylene glycol) methyl ether methacrylate (m-PEGMA) is often used to create PEG modified surfaces. Studies report the grafting of m-PEGMA on poly ether urethane surface by ozone induced polymerization, on emeraldine film by UV induced copolymerization.The methacrylate end-group allows for this material to be further polymerized.

Specifications

CAS 26915-72-0
Density 1.079 g/mL (at 25°C)
Refractive Index n20/D 1.4054
Linear Formula H2C=C(CH3)CO2(CH2CH2O)nCH3
Quantity 1 L
MDL Number MFCD00241432
Molecular Weight (g/mol) 2000
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.