Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(ethyleneimine) Solution, 50% (w/v) in Water, MilliporeSigma™ Supelco™

Catalog No. 111018445 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8445 1 L
11-102-0071 100 mL
11-101-8622 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-8445 Supplier MilliporeSigma Supelco Supplier No. P31431L
Add to Cart
Add to Cart

Specifications

CAS 9002-98-6
Concentration 50% (w/v) in H 2 O
Synonym Ethyleneimine polymer solution; PEI
Type Analytical Standard
Quantity 1 L
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.