Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly-L-Lysine Hydrobromide [MW 4,000-15,000], MP Biomedicals

Catalog No. ICN10269125 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10269125 25 mg
ICN10269180 100 mg
ICN10269190 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10269125 Supplier MP Biomedicals, Inc Supplier No. 0210269125
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 25988-63-0
Molecular Formula C18H38N6O4
Molecular Weight (g/mol) 402.54
InChI Key WBSCNDJQPKSPII-KKUMJFAQSA-N
Synonym lysyllysyllysine, l-lysyl-l-lysyl-l-lysine, unii-y7f82m80k8, trilysine, h-lys-lys-lys-oh, s-6-amino-2-s-6-amino-2-s-2,6-diaminohexanamido hexanamido hexanoic acid, l-lysine, l-lysyl-l-lysyl, 2olb, lysyl-lysyl-lysine, polylysine hbr
PubChem CID 72363
IUPAC Name (2S)-6-amino-2-[[(2S)-6-amino-2-[[(2S)-2,6-diaminohexanoyl]amino]hexanoyl]amino]hexanoic acid
SMILES C(CCN)CC(C(=O)NC(CCCCN)C(=O)NC(CCCCN)C(=O)O)N

Specifications

Quantity 25 mg
Formula Weight 4,000 to 15,000
Chemical Name or Material Poly-L-Lysine Hydrobromide
Recommended Storage 0°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.