Learn More
Advanced Biomatrix Poly-L-Ornithine, Non-Sterile Powder, 100 mg
Supplier:  Advanced Biomatrix 5172100MG
Encompass_Preferred
Poly-L-Ornithine comes with 100 mg of non-sterile powder. Poly-L-Ornithine is a synthetic amino acid chain that is positively charged and widely used as a coating to enhance cell attachment and adhesion to both plasticware and glass surfaces. The molecular weight of Poly-L-Ornithine can vary significantly with lower molecular weight (30,000 Da) being less viscous and higher molecular weight (>300,000 Da) having more binding sites per molecule. This product's molecular weight ranges from 70,000 to 150,000 Da. Poly-L-Ornithine is supplied in a 50 ml amber bottle. The product is ready-to-use after proper addition of cell culture water and sterile filtration.
Catalog No. NC2410839
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.