Learn More
Cell Signaling Technology Poly/Mono-ADP Ribose (D9P7Z) Rabbit mAb, High Specificity For ADP Ribosylated Proteins, Monoclonal Antibody, 100 µL
SDP
Supplier:  Cell Signaling Technology 89190S
Encompass
The Poly/Mono-ADP Ribose (D9P7Z) Rabbit Monoclonal Antibody is a highly specific tool for detecting ADP ribosylated proteins. Engineered for superior consistency and reliability, it is ideal for various applications such as Western Blotting, Immunofluorescence, and Flow Cytometry. This antibody uniquely recognizes endogenous levels of ADP ribosylated proteins without cross-reacting with other modifications, making it invaluable for research on cellular processes like DNA repair and cell stress response.
Catalog No. NC2822367
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.