Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Chromic Acid 10% Aqueous
SDP

Catalog No. 22023764
Change view
Click to view available options
Quantity:
16 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-023-764 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-023-764 Supplier StatLab Supplier No. S209916OZ
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

StatLab Poly Scientific Chromic Acid 10% Aqueous - used as a fixative to preserve cell structures

  • Chromic Acid 10% Aqueous is primarily used as a fixative to preserve cell structures and tissue samples for microscopic examination
  • The fixative function of chromic acid is based on its ability to cross-link proteins and other cellular components, resulting in the stabilization of cellular structures
  • A StatLab - Poly Scientific Special Stain Kit Component

Specifications

Chemical Name or Material Chromic Acid
Concentration 10%
Quantity 16 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.