Learn More
StatLab Poly Scientific Special Stain Kit Component, Hydrochloric Acid 20% Aqueous

Description
- A StatLab - Poly Scientific Special Stain Kit Component
- Hydrochloric Acid 20% Aqueous is used in Gomori's Method for Iron
Specifications
Specifications
| Physical Form | Aqueous Solution |
| Color | Clear Colorless |
| Quantity | 16 oz. |
| Grade | Laboratory |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.