Learn More
Pyramex Safety Products GL401 Polyurethane Coated Nylon Gloves
Shop All Pyramex Safety Products ProductsDescription
- 13 gauge nylon gloves with polyurethane coating
- Provides dexterity and tactile sensitivity
- Enhanced grip capabilities
- Low particulate shed
Specifications
Specifications
| ANSI Abrasion Rating | ANSI 1 |
| Cuff Material | Latex Elastic |
| Coating Material | Polyurethane |
| Color | Gray |
| Hand | Specific |
| Material | Nylon |
| Product Type | Seamless Knit Gloves |
| Size | Medium |
| EN388 Standard | 4131X |
| For Use With (Application) | Construction, Electronics, Inspection, and Quality Control |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.