Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex Safety Products GL402C5 Series Polyurethane Palm-Coated Gloves

Catalog No. 19555053 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Glove Cuff Color:
Black
Blue
Brown
Red
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Glove Cuff Color
19-555-053 Medium Yellow
19-555-054 Small Red
19-555-052 Large Brown
19-555-056 X-Large Black
19-555-055 2X-Large Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-555-053 Supplier Pyramex Safety Products Supplier No. GL402C5M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Polyurethane palm-coated gloves are tear and abrasion resistant, while offering tactile sensitivity and dexterity.

  • 13-gauge cut-resistant HPPE liner
  • Low particulate shed
  • Grips well without being sticky
  • Offers tactile sensitivity and dexterity
  • ANSI Cut Level: 4
  • Abrasion Rating: 4
  • Circular Blade Cut Rating: 5
  • Tear Rating: 4
  • Puncture Rating: 3

Specifications

ANSI Abrasion Rating ANSI 3
Coating Material Polyurethane
Color Gray, Green
Cut Resistance Rating ANSI A4
Material Polyurethane
Product Type Cut-resistant Gloves
Size Medium
EN388 Standard EN 388:4X43D
For Use With (Application) Electronics, general duty, parts assembly, sheet metal, handling, glass cutting
Glove Cuff Color Yellow
Glove Cuff Style Elasticated
Liner Material HPPE
Model GL402C5M
Puncture Rating ASTM F2878 LEVEL 3
Series GL402C5 Series Polyurethane Dipped
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.