Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(vinylidene fluoride)

Catalog No. p-7059151
Change view
Click to view available options
Quantity:
500 g
2 kg
10 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4408036 500 g
AA44080A3 2 kg
AA44080A9 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4408036 Supplier Thermo Scientific Chemicals Supplier No. 04408036
Only null left
Add to Cart

CAS: 24937-79-9 | (C2H2F2)n | NaN g/mol

Poly(vinylidene fluoride) is useful in coatings, film, filter cloth, instrument linings, filtration membranes, pump parts, linings and valves. It is also employed as a material for transducers in devices such as headphones, microphones and sonic detectors. It is also essential for piezoelectric and electrostrictive applications. In addition, it is useful for monofilament fishing lines replacing nylon monofilament. It acts as a standard binder used in the production of composite electrodes for lithium ion batteries. It serves as a high grade insulator for wires, nanomaterials and pressure sensors. It is utilized to make filters which are used to prepare samples for high performance liquid chromatography and other advanced analytical techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24937-79-9
Molecular Formula (C2H2F2)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00084470
InChI Key BQCIDUSAKPWEOX-UHFFFAOYSA-N
Synonym vinylidene fluoride, 1,1-difluoroethylene, ethene, 1,1-difluoro, vinylidene difluoride, pvdf, genetron 1132a, ethylene, 1,1-difluoro, halocarbon 1132a, poly vinylidene fluoride, kynar
PubChem CID 6369
ChEBI CHEBI:82550
IUPAC Name 1,1-difluoroethene
SMILES FC(F)(-*)C-*

Specifications

Color White
Physical Form Powder
Melting Point 155°C to 160°C
Solubility Information Soluble in organic esters,dimethylacetamide and amines. Insoluble in water.
Quantity 500 g
Odor Odorless
Chemical Name or Material Poly (vinylidene fluoride)
What is the average molecular weight of Poly(vinylidene fluoride)?

The average molecular weight of Poly(vinylidene fluoride) ranges from 250 and 450 kD.

TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.