Learn More
Sigma Organic Chemistry Polyethylene-block-poly(ethylene glycol) | 250G | MFCD00284335
Supplier: Sigma Organic Chemistry 458961250G

Polyethylene-block-poly(ethylene glycol), 250G
About This Item:
Linear Formula: CH3CH2(CH2CH2)X(OCH2CH2)YOH
MDL Number: MFCD00284335
UNSPSC Code: 12162002
Storage: room temp
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.