Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyanetholesulfonic Acid Sodium Salt, Reagent Grade, MP Biomedicals™

Catalog No. ICN10292305 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10292305 5 g
ICN10292325 25 g
ICN10292380 100 g
ICN10292390 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN10292305 Supplier MP Biomedicals, Inc Supplier No. 0210292305
Add to Cart
Add to Cart

Chemical Identifiers

CAS 52993-95-0
Molecular Formula (C10H11O4SNa)n
MDL Number MFCD00148427
Synonym sodium anethole, benzenesulfonic acid, methoxypropenyl-, sodium salt, homopolymer, sodium methoxypropenylbenzenesulonate homopolymer, benzenesulfonic acid, methoxy 1-propenyl-, sodium salt, homopolymer, sodium 1-methoxy-4-e-prop-1-enyl benzene, benzenesulfonic acid, methoxy 1-propen-1-yl-, sodium salt 1:1 , homopolymer

Specifications

Color White
Quantity 5 g
Formula Weight (250)n
Grade Reagent
Physical Form Solid
Chemical Name or Material Polyanetholesulfonic Acid Sodium Salt
Recommended Storage Store at room temperature.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.