Learn More
Promega PolyATtract™ mRNA Isolation System II with Magnetic Stand
Catalog No. PRZ5200 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
3 Isolations
Catalog No. Quantity
PRZ5200 3 Isolations
1 options
Catalog No. PRZ5200 Supplier Promega Supplier No. Z5200
Includes
  • Nuclease-Free Water
  • SSC, 20X
  • Biotinylated Oligo(dT) Probe
  • mRNA User Tube
  • MagneSphere™ Technology Magnetic Separation Stand (two-position)
  • Streptavidin MagneSphere™ Paramagnetic Particles
The PolyATtract System isolates messenger RNA directly from total RNA.
  • Isolates Highly Pure mRNA from Total RNA
  • Protocol complete in approximately 45 minutes
  • Isolated mRNA is suitable for use in in vitro translation, RT-PCR and cDNA synthesis
  • Cat.# Z5200 and Z5210 contain sufficient reagents for 3 separate mRNA isolations, each from 1-5mg total RNA
  • Cat.# Z5300 and Z5310 contain sufficient reagents for 15 separate mRNA isolations, each from 100-1,000μg total RNA
Specifications
Content And Storage 2°C to 10°C
Quantity 3 Isolations