Learn More
Promega PolyATtract™ mRNA Isolation Systems
Catalog No. PRZ5310 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
15 Preps
Includes:
mRNA Isolation System only
mRNA Isolation System, stand
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Includes
PR-Z5310 15 Preps mRNA Isolation System only
PR-Z5300 15 Preps mRNA Isolation System, stand
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PR-Z5310 Supplier Promega Supplier No. Z5310
The PolyATtract System isolates messenger RNA directly from total RNA.
  • Isolates Highly Pure mRNA from Total RNA
  • Protocol complete in approximately 45 minutes
  • Isolated mRNA is suitable for use in in vitro translation, RT-PCR and cDNA synthesis
  • Cat.# Z5200 and Z5210 contain sufficient reagents for 3 separate mRNA isolations, each from 1-5mg total RNA
  • Cat.# Z5300 and Z5310 contain sufficient reagents for 15 separate mRNA isolations, each from 100-1,000μg total RNA
Specifications
Content And Storage 2°C to 10°C; Nuclease-free Water 2 x 25mL, SSC (20X) 2 x 1.4mL, Biotinylated Oligo(dT) probe 1 x 50μL, Streptavidin MagneSphere™ paramagnetic particles 15 x 0.6mL
For Use With (Application) In-vitro translation, RT-PCR and cDNA synthesis
Includes mRNA Isolation System only
Quantity 15 Preps