Learn More
Promega PolyATtract™ System 1000 without Magnetic Stand
Catalog No. PRZ5400 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
Scalable
Catalog No. Quantity
PRZ5400 Scalable
1 options
Catalog No. PRZ5400 Supplier Promega Supplier No. Z5400
The PolyATtract System 1000 isolates messenger RNA directly from crude cell or tissue lysates, eliminating the need for total RNA isolation.
  • Isolates mRNA Directly from Crude Cell or Tissue Lysates
  • mRNA Purification is complete 45 minutes or less
  • Works with tissue amounts from 5mg-2g per isolation
  • No lengthy ethanol precipitation steps, phenol:chloroform extractions, and overnight ultracentrifugation
  • Magnetic separation stand (Cat.# Z5410) accommodates 1.5ml, 2ml, 15mL and 50mL tube sizes
Specifications
Content And Storage 2°C to 10°C
Quantity Scalable