Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Polycarbonate Test Tube Rack
SDPSCI_ED

Catalog No. S01241 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Positions:
12
18
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Positions
S01241 18
S01240 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S01241 Supplier United Scientific™ Supplier No. 77809
Add to Cart
Add to Cart
This item is not returnable. View return policy

Uniquely clear racks that allow you to see your sample.

These polycarbonate racks with a 3-tier design provide clear view of the tube contents and convenient insertion and removal of tubes. Upper and middle plates are transparent. Beveled holes on bottom plate provide stability to standing tubes. Racks are autoclavable and can withstand sub-freezing temperatures. Available in 12-tube and 18-tube configurations, each able to hold tubes up to 25 mm in diameter.

  • 3-tier design
  • Transparent plates for clear view of tubes
  • Holds tubes up to 25 mm in diameter

Specifications

Autoclavable Autoclavable
Color Clear/White
Holds Tubes (20 to 25 mm)
Material Polycarbonate
Positions 18
Shape Rectangular
Stackable No
Sterility Non-sterile
For Use With (Equipment) Test Tubes
Includes Test Tube Rack
Product Type Test Tube Rack
Shelves None
Array 3 x 6
Show More Show Less

Warning: Cancer and reproductive harm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.