Learn More
SETON SCIENTIFIC POLYCLEAR CENTRIFUGE TB 50/CS
Supplier: SETON SCIENTIFIC 7030
Open-Top Polyclear Tubes, Tube Size: 9/16 x 3-1/2 in., Tube Capacity: 12 mL, Temperature Range: > 25 deg. C, Can be used up to a pH of 8.5, Thin Wall and Translucent, Puncturable and Sliceable, Use with Cesium Chloride Gradients
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.