Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PolyCo™ PolyWear™ Smooth Gowns, Individually Bagged Bulk Packaged

Catalog No. 19125610
Change view
Click to view available options
Size:
Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-125-610 Large
19-125-611 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-125-610 Supplier PolyCo™ Supplier No. 10510
Add to Cart
Add to Cart
This item is not returnable. View return policy

Smooth finish gowns provide lightweight impervious protection against biohazards.

  • In compliance with FDA requirements and tested in accordance with ASTM standards including Breaking Load, Tensile Strength and Elongation
  • Successfully tested for liquid and aerosol blood resistance under ASTM F 1670 Synthetic Blood Penetration Resistance and ASTM F 1671 Bacteriophage Penetration Resistance
  • In compliance with OSHA Bloodborne Pathogen Standard, 29, CFR Part 1910.1030
  • Meets AAMI Levels 1-4
  • Superior physical properties over imports
  • Easy, tear-away feature for quick gown removal and safe disposal
  • Contoured thumb loops secure gown sleeve for added protection
  • Redirects stress to the film material not the thermal seals to minimize breakage potential
  • Full-length open back affords better comfort and shoulder coverage
  • Provides liquid and aerosol blood resistance
  • Lightweight, non-linting construction

Specifications

For Use With (Application) Hospitals, Health Clinics, Laboratories
Product Type General Purpose Gown
Size Large
Packaging Type Bulk, Individually Bagged
No. of Pockets None
Gender Unisex
Material Polyethylene Film
Color Blue
Closure Type Over-the-Head
Disposable Single-use
Sterility Non-sterile
Cuff Style Thumb Loop
Finish Smooth Matte
Latex Free Yes
Model 10510
Sleeve Style Full Sleeve with Thumb Loop
Certifications/Compliance ASTM F1670, ASTM F1671, AAMI Level 1 and 2, OSHA 29, CFR Part 1910.1030
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.