Learn More
PolyCo™ VR™ Die-Cut Aprons
Description
- Separate ties eliminated to avoid contamination problems
- Available in two lengths and three thicknesses
- Made in USA
Order Info
Standard or cleanroom packaged for Class 100 controlled environments.
Specifications
Specifications
| Material | Polyolefin |
| Color | Blue |
| Thickness | 6 mil |
| Dimensions (L x W) | 35 x 45 in. |