Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PolyCo™ VR™ Protective Boot Covers

Catalog No. 19125545
Change view
Click to view available options
Color:
Blue
Yellow
Size:
Large
Regular
Thickness:
4 mil
6 mil
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size Thickness
19-125-545 Yellow Regular 6 mil
19-125-543 Blue Regular 4 mil
19-125-544 Blue Large 4 mil
19-125-546 Yellow Large 6 mil
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-125-545 Supplier PolyCo™ Supplier No. 49355
Add to Cart
Add to Cart
This item is not returnable. View return policy

Provide knee-high, foot and lower leg protection in situations where biological and chemical liquids are present

  • Knee-high cover with elastic top
  • Two supple weights provide flexible, lightweight coverage
  • Available in different colors
  • Standard or cleanroom packaged (for Class 100 controlled environments)

Specifications

Size Regular
Material Polyolefin
Color Yellow
Closure Type Elastic Top
Seam Type Heat sealed
Thickness 6 mil
For Use With (Application) Ideal for use where biological and chemical liquids are present
Height (English) 14 in.
Height (Metric) 35.5 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.