Learn More
AdipoGen PolyD-lactide
SDP
Supplier:  AdipoGen CDXP0018M250
Encompass_Preferred
Chemical. CAS 106989-11-1. Formula C3H4O2n. MW 90000-124000. Synthetic. PolyD-lactide PDLA, a polymer of a stereospecific cyclic di-ester of lactic acid, is used in biomaterial research for the development of devices such as therapeutic drug delivery vessels. PolyD-lactide is used for the preparation of microparticles and resorbable polylactide scaffolds.
Catalog No. 50-256-0279
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.