Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 17,000

Catalog No. AA42492AK
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42492AK 250 mL
AA42492AD 50 mL
AA42492K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA42492AK Supplier Thermo Scientific Chemicals Supplier No. 042492AK
Add to Cart

CAS: 9016-00-6 | (C2H6OSi)n | 74.15 g/mol

Polydimethylsiloxane, trimethylsiloxy is used in Water-repellent, protective creams & lotions. It is also used as surfactant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9016-00-6
Molecular Formula (C2H6OSi)n
Molecular Weight (g/mol) 74.15
MDL Number MFCD00084411
InChI Key SEUDSDUUJXTXSV-UHFFFAOYSA-N
IUPAC Name dimethylsilanone
SMILES C[Si](C)(-*)O-*

Specifications

Color Colorless
Physical Form Viscous Liquid
Boiling Point 205°C
Melting Point <-40°C
Density 0.966 g/mL
Solubility Information Not miscible or difficult to mix with water. Insoluble in methanol,and ethylene glycol.
Quantity 250 mL
Flash Point 315°C (599°F)
Odor Odorless
Refractive Index 1.4033
Chemical Name or Material Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 17,000
Show More Show Less
RTECSNumber TQ2690000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.