Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Polyester Sign: DANGER: HOT. WILL CAUSE SEVERE BURNS OR INJURY.

Catalog No. 19110704 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W):
10 x 7 in.
14 x 10 in.
Displayed Text:
DANGER: HOT. WILL CAUSE SEVERE BURNS OR INJURY.
HOT. WILL CAUSE SEVERE BURNS OR INJURY.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Displayed Text
19-110-704 10 x 7 in. DANGER: HOT. WILL CAUSE SEVERE BURNS OR INJURY.
19-110-705 14 x 10 in. HOT. WILL CAUSE SEVERE BURNS OR INJURY.
19-110-726 10 x 7 in. HOT. WILL CAUSE SEVERE BURNS OR INJURY.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-110-704 Supplier Brady™ Supplier No. 143809

Improve workplace safety with proper signage and markings. This Brady™ polyester sign is a durable, clear and effective tool to raise awareness to workplace hazards.

  • Polyester material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use

Specifications

Material Polyester
Shape Rectangular
Dimensions (L x W) 10 x 7 in.
Displayed Text DANGER: HOT. WILL CAUSE SEVERE BURNS OR INJURY.
Color Black on White
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.